Copper(II) carbonate dihydroxide, Cu 55% min, Thermo Scientific Chemicals
Copper(II) carbonate dihydroxide, Cu 55% min, Thermo Scientific Chemicals
Copper(II) carbonate dihydroxide, Cu 55% min, Thermo Scientific Chemicals
Copper(II) carbonate dihydroxide, Cu 55% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(II) carbonate dihydroxide, Cu 55% min, Thermo Scientific Chemicals
CAS: 12069-69-1 | CH2Cu2O5
Have Questions?
Catalog NumberQuantity
ALF033305.36500 g
Catalog number ALF033305.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7440-67-7
IUPAC Namezirconium
Molecular FormulaZr
InChI KeyQCWXUUIWCKQGHC-UHFFFAOYSA-N
SMILES[Zr]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.5% (metals basis excluding Hf), Hf nominal 3%
Appearance (Color)Silvery
FormSlug, 6.35mm (0.25in) diameter.
Copper(II) carbonate dihydroxide is used as pigments and in treatment for copper deficiency in ruminants. It is also used in pyrotechnics, in sweetening of petrol sour crude stock and animal and poultry feeds. It finds application in some types of make-up such as lipstick. In analytical chemistry, it is used as a reagent for analysis.