Ammonium hydroxide, ACS, 28.0-30.0% NH{3}, Thermo Scientific Chemicals
Ammonium hydroxide, ACS, 28.0-30.0% NH{3}, Thermo Scientific Chemicals
Ammonium hydroxide, ACS, 28.0-30.0% NH{3}, Thermo Scientific Chemicals
Ammonium hydroxide, ACS, 28.0-30.0% NH{3}, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, ACS, 28.0-30.0% NH{3}, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number ALF033285.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium hydroxide
ColorColorless
CAS1336-21-6
Health Hazard 1H314-H335
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silve, and platinum jewelry. It is an active component of Tollens' reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.