Sulfurous acid, ACS, 6.0% SO{2} min, Thermo Scientific Chemicals
Sulfurous acid, ACS, 6.0% SO{2} min, Thermo Scientific Chemicals
Sulfurous acid, ACS, 6.0% SO{2} min, Thermo Scientific Chemicals
Sulfurous acid, ACS, 6.0% SO{2} min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfurous acid, ACS, 6.0% SO{2} min, Thermo Scientific Chemicals
CAS: 7782-99-2 | H2O3S | 82.073 g/mol
Have Questions?
Catalog NumberQuantity
ALF033275.AP500 mL
Catalog number ALF033275.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS543-59-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4105-1.4145 @ 20?C
Sulfurous acid is used as a reducing agent, bleaching agent and disinfectant. It is utilized for bleaching of moistened wool, straw for hats, and other materials injured by chlorine. It finds application as a disinfectant as well as used externally for its parasiticidal effect in various skin diseases. Its derivative viz. sulfites are included in antiseptics, antifermentatives, and antizymotics.