Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis), Thermo Scientific Chemicals
Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis), Thermo Scientific Chemicals
Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis), Thermo Scientific Chemicals
Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis), Thermo Scientific Chemicals
CAS: 5329-14-6 | H3NO3S | 97.088 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF033233.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS3029-19-4
IUPAC Namepyrene-1-carbaldehyde
Molecular FormulaC17H10O
InChI KeyRCYFOPUXRMOLQM-UHFFFAOYSA-N
SMILESO=CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark yellow to yellow
FormCrystals or powder or crystalline powder or lumps
Assay (HPLC)≥98.0%
Free acid (titration)≤1%
Melting Point (clear melt)123.0-130.0?C
Amidosulfonic acid is widely utilized as a green catalyst for the preparation of amide from ketoxime. It finds an application as a titrant in the determination of the burette injection volume and chemical calibration factor. It is an efficient catalyst in the synthesis of polyhydroquinoline derivatives by Hantzsch condensation reaction, deazaoxaflavin, fatty acid methyl and ethyl esters and in the isolation of 5-hydroxymethylfurfural (HMF) from bamboo fiber. It is involved in the determination of silicates in water samples. It acts as a neutralizing agent during the determination of paracetamol by Glynn and Kendal colorimetric method.