n-Octadecane, 99%, Thermo Scientific Chemicals
n-Octadecane, 99%, Thermo Scientific Chemicals
n-Octadecane, 99%, Thermo Scientific Chemicals
n-Octadecane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Octadecane, 99%, Thermo Scientific Chemicals
CAS: 593-45-3 | C18H38 | 254.502 g/mol
Have Questions?
Catalog NumberQuantity
ALF031954.22100 g
Catalog number ALF031954.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7681-38-1
IUPAC Namesodium hydrogen sulfate
Molecular FormulaHNaO4S
InChI KeyWBHQBSYUUJJSRZ-UHFFFAOYSA-M
SMILES[Na+].OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Titration with NaOH>=37.6 % (equivalent H2SO4)
Titration with NaOH>=90 %
Appearance (Form)Granules
Iron (Fe)=<200 ppm
View more
n-octadecane is used as solvent, lubricant, transformer oil and anti-corrosion agents. It is also used in paraffin as well as chemical intermediate in organic synthesis. It plays an important role in pheromones as a chemical messenger. N-Octadecane/ molecular sieve 5A composites find application as thermal storage material.