Trifluoroacetic acid, 99.5+%, Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5+%, Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5+%, Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5+%, Thermo Scientific Chemicals
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Catalog NumberQuantity
ALF031771.36500 g
Catalog number ALF031771.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTrifluoroacetic Acid
CAS76-05-1
Melting Point°C to 15°C
Boiling Point72°C to 73°C
Health Hazard 1H290-H314-H332-H335
View more
The combination of properties like solubility in most of the solvents, volatility, catalytic property and strong acidity with non-oxidizing nature makes it a widely used reagent in an organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and ceramic compounds. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. It finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.