n-Heptadecane, 99%, Thermo Scientific Chemicals
n-Heptadecane, 99%, Thermo Scientific Chemicals
n-Heptadecane, 99%, Thermo Scientific Chemicals
n-Heptadecane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptadecane, 99%, Thermo Scientific Chemicals
CAS: 629-78-7 | C17H36 | 240.475 g/mol
Have Questions?
Catalog NumberQuantity
ALF031551.1425 g
Catalog number ALF031551.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS603-35-0
IUPAC Nametriphenylphosphane
Molecular FormulaC18H15P
InChI KeyRIOQSEWOXXDEQQ-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
Melting point78.5°C to 81.5°C
GC>=98.5 %
n-Heptadecane is used as an internal standard to study the first direct comparison of kava lactone extraction efficiencies. It plays an important role as a suspended solvent used for the extraction and concentration of essential oil. It is used as a precursor in the production of iminoctadine triacetate