1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
Primary material for standardizing acids
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALF031395.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS1064-48-8
IUPAC Namedisodium (3Z)-5-amino-6-[2-(4-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC22H14N6Na2O9S2
InChI KeyHKBVRFLHNUEVRO-DWTBGCDMSA-L
SMILES[Na+].[Na+].NC1=C(N=NC2=CC=C(C=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N\NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Loss on Drying≤ 10.0%
UV/VisUV ratios: Absorption maxima wavelength: 617 to 620 nm
Assay (Dye)Dye Content: ≥ 84.00%
Appearance (Color)Dark blue to reddish-black powder
View more
Primary material for standardizing acids. Diphenylguanidine is used as a medium of natural and synthetic rubber accelerator. It acts as a complexing agent for the detection of metals and organic bases. Further, it is used as a secondary foam stabilizer in the silicoflouride foam process. It is actively involved in the manufacture of rubber sheet, rubber shoes and other rubber industrial products.