1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
CAS: 140-22-7 | C13H14N4O | 242.282 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number ALF031390.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS586-78-7
IUPAC Name1-bromo-4-nitrobenzene
Molecular FormulaC6H4BrNO2
InChI KeyZDFBKZUDCQQKAC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormPowder
Appearance (Color)White to cream to orange to brown
Identification (FTIR)Conforms
Melting Point (clear melt)122.0-128.0?C
1,5-Diphenylcarbazide is used in the colorimetric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.