1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
CAS: 140-22-7 | C13H14N4O | 242.282 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number ALF031390.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS124-41-4
IUPAC Namesodium methanolate
Molecular FormulaCH3NaO
InChI KeyWQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES[Na+].C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (non-U.S. sourced material)
Assay from Supplier's CofA≥97.5% (U.S. sourced material)
Identification (FTIR)Conforms (non-U.S. sourced material)
FormPowder
CommentMaterial sourced in the U.S. and in other countries
View more
1,5-Diphenylcarbazide is used in the colorimetric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.