1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
CAS: 14634-91-4 | C36H24FeN6O4S | 692.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF030562.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS937-14-4
IUPAC Name3-chlorobenzene-1-carboperoxoic acid
Molecular FormulaC7H5ClO3
InChI KeyNHQDETIJWKXCTC-UHFFFAOYSA-N
SMILESOOC(=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Moist powder
3-Chlorobenzoic acid=<12 %
HPLCdi-m-Chlorobenzoyl peroxide: < 0.1% Typical
HPLC=<0.2 % di-m-Chlorobenzoyl peroxide
Titration Iodometric70 to 75 %
View more
1,10-Phenanthroline iron(II) sulfate is used as an indicator for the iron phenanthroline compound in analytical chemistry. Further, it is used as a redox indicator for making oscillatory Belousov-Zhabotinsky reactions visible.