Trimethylphosphine oxide, Thermo Scientific Chemicals
Trimethylphosphine oxide, Thermo Scientific Chemicals
Trimethylphosphine oxide, Thermo Scientific Chemicals
Trimethylphosphine oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trimethylphosphine oxide, Thermo Scientific Chemicals
CAS: 676-96-0 | C3H9OP | 92.078 g/mol
Have Questions?
Catalog NumberQuantity
ALF030394.065 g
Catalog number ALF030394.06
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS94790-37-1
IUPAC Name(1H-1,2,3-benzotriazol-1-yl)[bis(dimethylamino)methylidene]oxidanium
Molecular FormulaC11H16N5O
InChI KeyCLZISMQKJZCZDN-UHFFFAOYSA-N
SMILESCN(C)C(=[O+]N1N=NC2=CC=CC=C12)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystalline powder
Assay (HPLC)≥97.5%
Loss on Drying<1.0%
Appearance (Color)White to pale cream
Trimethylphosphine oxide is a common side product in organic synthesis, including the Wittig, Staudinger, Mitsunobu reactions and also in the manufacture of triphenylphosphine. It is a popular reagent to induce the crystallization of various chemical compounds. It acts as an excellent ligand for hard metal centers. It can react with 9-methyl-9-azonianthracene methyl sulfate to get (10-methyl-9,10-dihydro-acridin-9-yl)-phosphonic acid dimethyl ester using sodium iodide and acetonitrile as solvent.