Formaldehyde, Quant™ Test Strips
Formaldehyde, Quant™ Test Strips
Formaldehyde, Quant™ Test Strips
Have Questions?
Catalog NumberQuantity
ALF023277.QA1 Kit
Catalog number ALF023277.QA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Kit
Request bulk or custom format
Chemical Identifiers
CAS2389-45-9
IUPAC Name(2S)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Molecular FormulaC19H28N2O6
InChI KeyBDHUTRNYBGWPBL-GGYSOQFKNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%