Chloride Quant™ Test Strips
Chloride Quant™ Test Strips
Chloride Quant™ Test Strips
Have Questions?
Catalog number ALF023271.QA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS67533-12-4
IUPAC Nametetraethylazanium acetate tetrahydrate
Molecular FormulaC10H31NO6
InChI KeyQNOAKQGNSWGYOE-UHFFFAOYSA-M
SMILESO.O.O.O.CC([O-])=O.CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point41°C to 46°C
Titration with HClO497.0 to 103.0 %