2-Butanone, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
2-Butanone, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
2-Butanone, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
2-Butanone, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Butanone, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
CAS: 78-93-3 | C4H8O | 72.11 g/mol
Have Questions?
Catalog NumberQuantity
ALF022924.K21 L
Catalog number ALF022924.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS645-08-9
IUPAC Name3-hydroxy-4-methoxybenzoate
Molecular FormulaC8H7O4
InChI KeyLBKFGYZQBSGRHY-UHFFFAOYSA-M
SMILESCOC1=CC=C(C=C1O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (DSC)246.0-255.0°C
FormPowder
Appearance (Color)White to cream to pale brown
Assay (Aqueous acid-base Titration)≥96.5 to ≤104.5%
Water Content (Karl Fischer Titration)≤0.5%
View more
2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO. It is used as a mobile phase in High Performance Liquid Chromatography.