1,4-Dioxane, HPLC Grade, 99% min, Thermo Scientific Chemicals
1,4-Dioxane, HPLC Grade, 99% min, Thermo Scientific Chemicals
1,4-Dioxane, HPLC Grade, 99% min, Thermo Scientific Chemicals
1,4-Dioxane, HPLC Grade, 99% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, HPLC Grade, 99% min, Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Catalog NumberQuantity
ALF022913.K21 L
Catalog number ALF022913.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7772-99-8
IUPAC Nameλ²-tin(2+) dichloride
Molecular FormulaCl2Sn
InChI KeyAXZWODMDQAVCJE-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Iron (Fe)=<50 ppm
Appearance (Form)Crystalline powder or flakes
Lead (Pb)=<75 ppm
Titration Iodimetric97.5 to 102.5 %
1,4-Dioxane HPLC Grade is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.