Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Chelating agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ALF018697.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Chemical Identifiers
CAS78-78-4
IUPAC Name2-methylbutane
Molecular FormulaC5H12
InChI KeyQWTDNUCVQCZILF-UHFFFAOYSA-N
SMILESCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 240 nm A: =<0.01
UVat 210 nm A: =<0.30
UV1 cm cell vs HPLC-grade water
UVat 220 nm A: =<0.07
Color scale=<10 APHA
View more
Zinc diethyldithiocarbamate is a chelating ligand. It is used as a common accelerator to latex. A fast curing primary or secondary effective ultra-accelerator for natural and synthetic latex compounds.