Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Chelating agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number ALF018697.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZinc diethyldithiocarbamate
CAS14324-55-1
Health Hazard 1H302-H315-H317-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
View more
Zinc diethyldithiocarbamate is a chelating ligand. It is used as a common accelerator to latex. A fast curing primary or secondary effective ultra-accelerator for natural and synthetic latex compounds.