Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT, Thermo Scientific Chemicals
Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT, Thermo Scientific Chemicals
Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT, Thermo Scientific Chemicals
Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT, Thermo Scientific Chemicals
CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Catalog NumberQuantity
ALF016767.36500 g
Catalog number ALF016767.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS93-07-2
IUPAC Name3,4-dimethoxybenzoic acid
Molecular FormulaC9H10O4
InChI KeyDAUAQNGYDSHRET-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1OC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥99.0 to ≤101.0%
Melting Point (clear melt)179.0-186.0?C
FormPowder
Assay (Silylated GC)≥99.0%
Solvent for U.V. spectrophotometric and it is also important as solvent in production of cellulose plastics such as cellulose acetate. It is also common solvent for Grignard reaction.