Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF016198.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7789-09-5
IUPAC Namediammonium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2H8N2O7
InChI KeyJOSWYUNQBRPBDN-UHFFFAOYSA-P
SMILES[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Appearance (Form)Adhering crystals
Titration Iodometric>=98.5 %
Sulfate (SO4)=<500 ppm
Loss on drying=<3 % (105°C, 4 h)
View more
As a reagent for ion-pair extractionsTetra-n-butylammonium hydroxide, 55% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.