Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALF016198.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS7251-61-8
IUPAC Name2-methylquinoxaline
Molecular FormulaC9H8N2
InChI KeyALHUXMDEZNLFTA-UHFFFAOYSA-N
SMILESCC1=CN=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear orange to dark red
Appearance (Form)Liquid
AppearanceMay darken on storage
GC>=96.0 %
As a reagent for ion-pair extractionsTetra-n-butylammonium hydroxide, 55% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.