Iron powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
Iron powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
Iron powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
Iron powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iron powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF014183.30250 g
Catalog number ALF014183.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIron powder
CAS7439-89-6
Health Hazard 1H500
Recommended StorageAmbient temperatures
EINECS Number231-096-4