Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals
Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals
Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals
Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals
CAS: 6487-48-5 | C2H2K2O5 | 184.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF013452.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS64424-12-0
IUPAC Namelanthanum(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21LaO6
InChI KeyUUYNRMJIFVOQCR-UHFFFAOYSA-N
SMILES[La+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Potassium oxalate monohydrate is used as a chelating agent; a purifying agent in pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, a rust remover for metal treatment, a bleaching agent in textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as a reducing agent in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.