Gallium Indium eutectic, 99.99% (metals basis), Thermo Scientific Chemicals
Gallium Indium eutectic, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallium Indium eutectic, 99.99% (metals basis), Thermo Scientific Chemicals
Gallium Indium eutectic, Ga:In; 75.5:24.5 wt%, 50 g, 99.99% (metals basis), 6.2, Hazard Class: 8; Packaging Group: III, 231-163-8, H290-H314-H335, P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c, 15.7 deg.C, MFCD00144387, Liquid
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Catalog number ALF012478.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1614-12-6
IUPAC Name1H-1,2,3-benzotriazol-1-amine
Molecular FormulaC6H6N4
InChI KeyJCXKHYLLVKZPKE-UHFFFAOYSA-N
SMILESNN1N=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point81°C to 85°C
HPLC>=97.5 %
It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices. Gallium-Indium eutectic top contact, and a halogen light source was used to test P3HT-PCBM organic solar cells.