Platinum black, Thermo Scientific Chemicals
Platinum black, Thermo Scientific Chemicals
Platinum black, Thermo Scientific Chemicals
Platinum black, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum black, Thermo Scientific Chemicals
Form: S.A. 10m2/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALF012076.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS79-00-5
IUPAC Name1,1,2-trichloroethane
Molecular FormulaC2H3Cl3
InChI KeyUBOXGVDOUJQMTN-UHFFFAOYSA-N
SMILESClCC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4700 to 1.4720 (20°C, 589 nm)
GC>=97.5 %
Platinum black is used in polymer electrolyte fuel cells, microbial fuel cells, and phosphoric acid fuel cell applications. It is also used as a recombination catalyst for hydrogen elimination. It is also used as a building block for sensor elements. It is used as a catalyst and as a gas absorbent. It is also useful for the electrochemical detection of hydrogen peroxide and nitrite in microchannel. It is useful for the anodic oxidation of ammonia in aqueous potassium hydroxide electrolyte.