ROTAVISC lo-vi HELI Complete Viscometer Package
ROTAVISC lo-vi HELI Complete Viscometer Package
Actual product may vary
ROTAVISC lo-vi HELI Complete Viscometer Package
ROTAVISC lo-vi HELI Complete Viscometer Package
Thermo Scientific Chemicals
ROTAVISC lo-vi HELI Complete Viscometer Package
The ROTAVISC lo-vi HELI Complete Viscometer Package determines the viscosity of low-viscosity liquids in all areas of application ranging from the laboratory to quality control.
Have Questions?
Catalog number ALF011831.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Package includes a standard spindle set (SP 1-SP 4), a protective bracket, temperature sensor and HELISTAND stand with T-spindle set
  • Easy to operate
  • Large 4.3 in. TFT display allows for intuitive menu guidance
  • Simple ramp function simplifies repetitive tasks
  • High measurement accuracy for both Newtonian and non-Newtonian fluids of ±1% of the measuring range
  • Reproducibility within ±0.2%
  • Stepless speed adjustment
  • Motorized IKA™ HELISTAND moves the mounted viscometer and its rotating body slowly and uniformly up and down in the sample
Specifications
Product TypeRotational Viscometer
Viscosity Range15 to 6,000,000 cSt
Display TypeTFT
Certifications/ComplianceCE
IncludesROTAVISC lo-vi HELI complete viscometer, Spindle Set-1, PT 100.8 temperature probe, HELISTAND motorized stand and T-Spindle-Set
Height (English)24.8 in.
Height (Metric)629 mm
Length (English)14.6 in.
Length (Metric)372 mm
Width (English)13.8 in.
Width (Metric)351 mm
Accuracy0.01
Repeatability0.20%
Count (Speed Setting)Stepless
For Use With (Application)Viscosity measurement of flowing and non-flowing liquids
SensorsTemperature Sensor
Speed Range0.01 to 200 rpm
Temperature Range (Metric)-100°C to +300°C
Voltage100/240 V
Unit SizeEA