Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF011067.30250 g
Catalog number ALF011067.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS92-77-3
IUPAC Name3-hydroxy-N-phenylnaphthalene-2-carboxamide
Molecular FormulaC17H13NO2
InChI KeyJFGQHAHJWJBOPD-UHFFFAOYSA-N
SMILESOC1=C(C=C2C=CC=CC2=C1)C(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration excess NaOH>=98.5 %
Appearance (Form)Powder
Appearance (Color)Beige
Melting point>=243°C