Check Order Status
Long Ground Cones With Tip
Thermo Scientific Chemicals
Long Ground Cones With Tip
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Have Questions?
Catalog Number
Joint Size
ALF011008.14
NS 14/35
Catalog number
ALF011008.14
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Joint Size:
NS 14/35
Conical ground cones
Made of DURAN™ tubing
Specifications
Quantity
10/Pk.
Joint Size
NS 14/35
Thickness (Metric) Wall
1.5 mm
Diameter (Metric) Outer Tube
13 mm
Length (Metric)
120 mm
Material
Borosilicate Glass (3.3)
Unit Size