Niobium(V) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Niobium(V) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Niobium(V) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Niobium(V) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Niobium(V) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
CAS: 1313-96-8 | Nb2O5 | 265.81 g/mol
Have Questions?
Catalog NumberQuantity
ALF010822.1850 g
Catalog number ALF010822.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS89-83-8
IUPAC Name5-methyl-2-(propan-2-yl)phenol
Molecular FormulaC10H14O
InChI KeyMGSRCZKZVOBKFT-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point47.5-52.5?C
Appearance (Color)White
Identification (FTIR)Conforms
Niobium pentoxide is the precursor to a majority of niobium compounds, powder metallurgy products, and alloys. It is also used in glass, optic and ceramic applications. The nanoparticles of niobium pentoxide are useful in the preparation of electrochemical capacitors and cathodes. In addition, appropriate preparations of this compound can be used as a dietary supplement in food industry. It is a precursor to lithium niobate which finds extensive use in piezoelectric sensors, anti-aliasing filters, optical waveguides, mobile phones, surface acoustic devices, optical modulators, including linear and non-linear optical applications.