Cobalt powder, -60 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Cobalt powder, -60 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Cobalt powder, -60 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Cobalt powder, -60 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt powder, -60 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF010456.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCobalt
ColorBlack
CAS7440-48-4
Health Hazard 1H228-H302-H317-H319-H330-H334-H335-H336-H350i-H360-H373-H500
Health Hazard 3P201-P202-P210-P240-P241-P260-P264b-P270-P271-P272-P280-P281-P284-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P370+P378q-P501c