Sodium, 99.95% (metals basis), Thermo Scientific Chemicals
Sodium, 99.95% (metals basis), Thermo Scientific Chemicals
Sodium, 99.95% (metals basis), Thermo Scientific Chemicals
Sodium, 99.95% (metals basis), Thermo Scientific Chemicals
Sodium, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7440-23-5 | Na | 22.99 g/mol
Have Questions?
Catalog NumberQuantity
ALF010336.065 g
Catalog number ALF010336.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS115-22-0
IUPAC Name3-hydroxy-3-methylbutan-2-one
Molecular FormulaC5H10O2
InChI KeyBNDRWEVUODOUDW-UHFFFAOYSA-N
SMILESCC(=O)C(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to light red
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4140 to 1.4170 (20°C, 589 nm)
GC>=90 %