Potassium, solid, 99.5% (metals basis), Thermo Scientific Chemicals
Potassium, solid, 99.5% (metals basis), Thermo Scientific Chemicals
Potassium, solid, 99.5% (metals basis), Thermo Scientific Chemicals
Potassium, solid, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium, solid, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-09-7 | K | 39.10 g/mol
Have Questions?
Catalog NumberQuantity
ALF010299.031 g
Catalog number ALF010299.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS79-81-2
IUPAC Name(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl hexadecanoate
Molecular FormulaC36H60O2
InChI KeyVYGQUTWHTHXGQB-FFHKNEKCSA-N
SMILESCCCCCCCCCCCCCCCC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to brown
FormLiquid or oil
Assay (unspecified)≥1.7 MIU/g
Assay from Supplier's CofA≥94.0% (HPLC)