Iron powder, spherical, APS 6-10 micron, reduced, 99.5%, Thermo Scientific Chemicals
Iron powder, spherical, APS 6-10 micron, reduced, 99.5%, Thermo Scientific Chemicals
Iron powder, spherical, APS 6-10 micron, reduced, 99.5%, Thermo Scientific Chemicals
Iron powder, spherical, APS 6-10 micron, reduced, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iron powder, spherical, APS 6-10 micron, reduced, 99.5%, Thermo Scientific Chemicals
Apparent density 2.0-3.0g/cm3
Have Questions?
Catalog NumberQuantity
ALF010214.1425 g
Catalog number ALF010214.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2079-89-2
IUPAC Name(2E)-but-2-enedioic acid; bis(3-aminopropanenitrile)
Molecular FormulaC10H16N4O4
InChI KeyNYMXYZMHOZAPHQ-SEPHDYHBSA-N
SMILESNCCC#N.NCCC#N.OC(=O)\C=C\C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or granules
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Appearance (Color)White to pale cream