CERILLIANT™ Phenobarbital-D5 (side chain) Solution, (deuterium label on side chain), Sold by MilliporeSigma™ Supelco™
CERILLIANT™ Phenobarbital-D5 (side chain) Solution, (deuterium label on side chain), Sold by MilliporeSigma™ Supelco™
CERILLIANT™ Phenobarbital-D5 (side chain) Solution, (deuterium label on side chain), Sold by MilliporeSigma™ Supelco™
CERILLIANT™ Phenobarbital-D5 (side chain) Solution, (deuterium label on side chain), Sold by MilliporeSigma™ Supelco™
Thermo Scientific Chemicals
CERILLIANT™ Phenobarbital-D5 (side chain) Solution, (deuterium label on side chain), Sold by MilliporeSigma™ Supelco™
Certified reference material
Have Questions?
Catalog NumberQuantity
ALF010018.221 mL
Catalog number ALF010018.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 mL
Request bulk or custom format
Chemical Identifiers
CAS67-56-1, 73738-05-3
Specifications
CAS67-56-1
Health Hazard 1H311
Health Hazard 2H331-H370
CAS73738-05-3
Recommended Storage2 to 8°C
View more
An internal standard suitable for quantitation of phenobarbital levels in GC/MS or LC/MS applications for clinical toxicology, urine drug testing, or forensic analysis. Phenobarbital is a barbiturate used to treat all epileptic seizure types except for absence seizures. Phenobarbital is the oldest and most widely used anticonvulsant.
CERILLIANT is a registered trademark of Cerilliant Corporation