Magnesium granules, -4+30 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
Magnesium granules, -4+30 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
Magnesium granules, -4+30 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
Magnesium granules, -4+30 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium granules, -4+30 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Catalog number ALF000870.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialMagnesium granules
ColorSilver to Gray
CAS7439-95-4
Health Hazard 1H228-H252-H261
Health Hazard 2GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.