Germanium pieces, 2cm (0.8in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals
Germanium pieces, 2cm (0.8in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals
Germanium pieces, 2cm (0.8in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals
Germanium pieces, 2cm (0.8in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Germanium pieces, 2cm (0.8in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals
CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Catalog NumberQuantity
ALF000123.0910 g
Catalog number ALF000123.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS865-48-5
IUPAC Namesodium 2-methylpropan-2-olate
Molecular FormulaC4H9NaO
InChI KeyMFRIHAYPQRLWNB-UHFFFAOYSA-N
SMILES[Na+].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
NaOH + Na2CO3 (as NaOH)=<1.5 %
Titration with HCl98.5 to 101.5 %