Aluminum foil, 0.13mm (0.005in) thick, 50x125mm (2.0x4.9in), 99.9995% (metals basis), Thermo Scientific Chemicals
Aluminum foil, 0.13mm (0.005in) thick, 50x125mm (2.0x4.9in), 99.9995% (metals basis), Thermo Scientific Chemicals
Aluminum foil, 0.13mm (0.005in) thick, 50x125mm (2.0x4.9in), 99.9995% (metals basis), Thermo Scientific Chemicals
Aluminum foil, 0.13mm (0.005in) thick, 50x125mm (2.0x4.9in), 99.9995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum foil, 0.13mm (0.005in) thick, 50x125mm (2.0x4.9in), 99.9995% (metals basis), Thermo Scientific Chemicals
≈2.2g/50x125mm
Have Questions?
Catalog NumberQuantity
ALF000004.KF1 Ea.
Catalog number ALF000004.KF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Ea.
Request bulk or custom format
Chemical Identifiers
CAS19764-30-8
IUPAC Name(2R)-2-acetamido-4-methylpentanoic acid
Molecular FormulaC8H15NO3
InChI KeyWXNXCEHXYPACJF-JLDDOWRYNA-N
SMILESCC(C)C[C@@H](NC(C)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)176.0-184.0?C
Optical Rotation24.0 ± 2.0? (C=5 in ethanol)