Check Order Status
Silica Gel Tel-Tale Desiccant, Puricore™
Element Search
Structure Search
Thermo Scientific Chemicals
Silica Gel Tel-Tale Desiccant, Puricore™
Have Questions?
Catalog Number
Quantity
ACRUSS161212
2.5 kg
Catalog number
ACRUSS161212
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS
7553-56-2
IUPAC Name
diiodine
Molecular Formula
I2
InChI Key
PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES
II
View more
Specifications
Specification Sheet
Insoluble matter
Passes test
Appearance (Form)
Metallic spheres
Titration Iodometric
>=99.4 %
Chloride and Bromide
=<0.005 %
Appearance (Color)
Dark grey