Phosphoric acid, for HPLC, Thermo Scientific Chemicals
Phosphoric acid, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, for HPLC, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ACR463865000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPhosphoric acid
CAS7664-38-2
Melting Point42.0°C
Health Hazard 1GHS Signal Word: Danger
Density1.7000 g/mL