Lovastatin, 98%
Lovastatin, 98%
Lovastatin, 98%
Thermo Scientific Chemicals
Lovastatin, 98%
Lovastatin, CAS # 75330-75-5, is a cholesterol lowering compound and inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ACR458830050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS103-23-1
IUPAC Name1,6-bis(2-ethylhexyl) hexanedioate
Molecular FormulaC22H42O4
InChI KeySAOKZLXYCUGLFA-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4440 to 1.4500 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<30 APHA
Infrared spectrumConforms
GC>=98.5 %
General Description
Applications
• It blocks product ion of mevalonate, a critical compound in the production of cholesterol and isoprenoids
• It can increase cellular resistance to anticancer agents such as doxorubicin • It can induce apoptosis in myeloma cells
• It can cause cell cycle arrest in G1 and G2/M phase
• It also blocks PDGF receptor tyrosine phosphorylation and MAP kinase activation by PDGF
RUO – Research Use Only
General References:
  1. Tobert, A.J. Efficacy and long-term adverse effect pattern of lovastatin. The American Journal of Cardiology. 1988, 62, (15),J28-J34.
  2. Jones, P. MD; Kafonek, S. MD; Laurora, I. PharmD; Donald Hunninghake MD. Comparative Dose Efficacy Study of Atorvastatin VersusSimvastatin, Pravastatin, Lovastatin, and Fluvastatin in Patients With Hypercholesterolemia (The CURVES Study). The American Journal of Cardiology. 1998, 81, (5),582-587.