Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number ACR458690025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS5597-50-2
IUPAC Namemethyl 3-(4-hydroxyphenyl)propanoate
Molecular FormulaC10H12O3
InChI KeyXRAMJHXWXCMGJM-UHFFFAOYSA-N
SMILESCOC(=O)CCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%