Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number ACR458690010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS98-00-0
IUPAC Name(furan-2-yl)methanol
Molecular FormulaC5H6O2
InChI KeyXPFVYQJUAUNWIW-UHFFFAOYSA-N
SMILESOCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
AppearanceMay darken on storage
Appearance (Form)Liquid
GC>=97.5 %