Ammonium hydroxide, Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number ACR458680025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)minimum 99.5%
Particle SizeAPS 6-10 micron