Ammonium hydroxide, Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number ACR458680010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS16971-33-8
IUPAC Namechloro(hydrido)rutheniumiumylium; methanidylidyneoxidanium; tris(triphenylphosphane)
Molecular FormulaC55H46ClOP3Ru
InChI KeyIMPHYUNMDWWJAH-UHFFFAOYSA-M
SMILESCl[RuH++].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to tan to brown
Ruthenium (Ru)>=10.3 %
Appearance (Form)Powder or crystals