Furan-2-acetic acid, 97%, Thermo Scientific Chemicals
Furan-2-acetic acid, 97%, Thermo Scientific Chemicals
Furan-2-acetic acid, 97%, Thermo Scientific Chemicals
Furan-2-acetic acid, 97%, Thermo Scientific Chemicals
Acros Organics
Furan-2-acetic acid, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR458390010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS66648-50-8
IUPAC Nameethyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
Molecular FormulaC11H12O4
InChI KeyWDKYDMULARNCIS-GQCTYLIASA-N
SMILESCCOC(=O)C=CC1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormPowder
Melting Point (clear melt)142.0-148.0?C
Assay (GC)≥96.0%