3-Phenylpropyl isocyanate, 97%, Thermo Scientific™
3-Phenylpropyl isocyanate, 97%, Thermo Scientific™
3-Phenylpropyl isocyanate, 97%, Thermo Scientific™
3-Phenylpropyl isocyanate, 97%, Thermo Scientific™
Acros Organics
3-Phenylpropyl isocyanate, 97%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR457380050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Phenylpropyl isocyanate
CAS68664-23-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritatio
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortab