Oxaliplatin, 99%
Oxaliplatin, 99%
Oxaliplatin, 99%
Thermo Scientific Chemicals
Oxaliplatin, 99%
CAS: 61825-94-3 | C8H14N2O4Pt | 397.29 g/mol
Have Questions?
Catalog NumberQuantity
ACR456131000100 mg
Catalog number ACR456131000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS103-47-9
IUPAC Namesodium 2-(cyclohexylamino)ethane-1-sulfonate
Molecular FormulaC8H16NNaO3S
InChI KeyTXWKJOVDGFRVOG-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CCNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤0.5%
Appearance (Color)White