Arbutin, 98%, Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
CAS: 497-76-7 | C12H16O7 | 272.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ACR455640250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialArbutin
CAS497-76-7
Melting Point195.0°C to 201.0°C
EINECS Number207-850-3
Loss on Drying2% max.