Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
CAS: 60-35-5 | C2H5NO | 59.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number ACR451872500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.998% (metals basis)
FormFoil, 2.0mm (0.08in) thick.