Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
CAS: 60-35-5 | C2H5NO | 59.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ACR451870010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS1066-35-9
IUPAC Namechlorodimethylsilyl
Molecular FormulaC2H6ClSi
InChI KeyQABCGOSYZHCPGN-UHFFFAOYSA-N
SMILESC[Si](C)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear to slightly hazy colorless to pale yellow
CommentHaziness may settle as precipitate on storage but will not affect performance